Learn More
Hach Company Buffer Powder Pillows, pH 8.00, pk/15
Supplier:  Hach Company 1407995
Encompass
pH Buffer Powder Pillows for pH electrode calibration. Each powder Pillow prepares 50 mL of fresh buffer solution. Colorless. pH is 8.00 ±0.02 at 25°C. Pack of 15 Powder Pillows. Pack of 15 Powder Pillows
Catalog No. 50-199-4921