Learn More
Aqua Solutions BUFFER REF PH 11.0 (+-0.01) 1L
SDP
Catalog No. NC0366467
Encompass_Preferred
BUFFER REFERENCE pH 11.0 (+- 0.01 @ 25C) N.I.S.T. TRACEABLE PREPARED TO ASTM D 664
Catalog No. NC0366467 Supplier Aqua Solutions Supplier No. 15281L
BUFFER REFERENCE pH 11.0 (+- 0.01 @ 25C) N.I.S.T. TRACEABLE PREPARED TO ASTM D 664