Learn More
Aqua Solutions BUFFER REFERENCE PH2.0 500ML
SDP
Catalog No. NC9304863
Encompass_Preferred
Buffer solution, pH2, Traceable to N.I.S.T., 500mL
Catalog No. NC9304863 Supplier Aqua Solutions Supplier No. 1440500ML
Buffer solution, pH2, Traceable to N.I.S.T., 500mL