Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer, Reference Standard, pH 10.00 ± 0.01 at 25°C, Ricca Chemical
SDP

Catalog No. 16001 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Natural Poly Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
1600-1 4 L Natural Poly Bottle
1600-16 500 mL Natural Poly Bottle
1600-32 1 L Natural Poly Bottle
1600-1G 4 L Amber Glass Bottle
16001CT 4 L Cubitainer
1600-25 10 L Cubitainer
1600-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 1600-1 Supplier Ricca Chemical Company Supplier No. 16001
Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 10.00 ± 0.01 at 25 deg C
CAS 497-19-8, 144-55-8, 1310-73-2, 7732-18-5
CAS Min % 97.0, 0.51, 0.39, 0.03
CAS Max % 100.0, 0.53, 0.41, 0.03
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 10
Physical Form Liquid
Quantity 4 L
Solution Type pH Buffer
Packaging Natural Poly Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.