Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer, Reference Standard, pH 10.00 ± 0.01 at 25°C (Color Coded Blue), Ricca Chemical
GSA_VASDP

Catalog No. 16015 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
10 L
20 L
200 x 20 mL
Packaging:
Cubitainer
Natural Poly Bottle
Pack
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
1601-5 20 L Cubitainer
160120 20 x 20 mL Pack
1601-16 500 mL Natural Poly Bottle
1601-32 1 L Natural Poly Bottle
1601-1CT 4 L Cubitainer
1601-1 4 L Natural Poly Bottle
16-012-00 200 x 20 mL Pack
160125 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 1601-5 Supplier Ricca Chemical Company Supplier No. 16015
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 10.00 ± 0.01 at 25 deg C (Color Coded Blue)
Melting Point 0°C
CAS 497-19-8, 144-55-8, 7732-18-5
Boiling Point 100°C
CAS Min % 97.0, 0.51, 0.39
Color Blue
CAS Max % 100.0, 0.53, 0.41
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 10
Physical Form Liquid
Quantity 20 L
Solution Type Colored pH Buffer
Packaging Cubitainer
Solubility Information Miscible
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.