Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer, Reference Standard, pH 4.63 ± 0.01 at 25°C, Ricca Chemical
SDP

Catalog No. 150416 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1504-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1504-16 Supplier Ricca Chemical Company Supplier No. 150416
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference standard buffer for general laboratory use.

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 4.63± 0.01 at 25°C
CAS 877-24-7, 1310-73-2, 7732-18-5
CAS Min % 97.0, 0.99, 0.07
Color Colorless
CAS Max % 100.0, 1.03, 0.07
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 4.63
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Packaging Natural Poly Bottle
Grade Not Available
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.