Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer, Reference Standard, pH 7.00 ± 0.01 at 25°C (Color Coded Yellow), Ricca Chemical
SDP

Catalog No. 15511G Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
20 mL
500 mL
1 L
4 L
10 L
20 L
200 x 20 mL
Packaging:
Amber Glass Bottle
Cubitainer
Natural Poly Bottle
Pack
Pouch
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
1551-1G 4 L Amber Glass Bottle
155120 20 mL Pouch
1551-16 500 mL Natural Poly Bottle
1551-32 1 L Natural Poly Bottle
15511CT 4 L Cubitainer
1551-1 4 L Natural Poly Bottle
15-512-00 200 x 20 mL Pack
1551-25 10 L Cubitainer
1551-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 1551-1G Supplier Ricca Chemical Company Supplier No. 15511G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 7.00± 0.01 at 25°C (Color Coded Yellow)
CAS 7558-79-4, 7778-77-0, 7732-18-5
CAS Min % 97.0, 0.87, 0.48
Color Yellow
CAS Max % 100.0, 0.91, 0.5
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 7
Physical Form Liquid
Quantity 4 L
Solution Type Colored pH Buffer
Packaging Amber Glass Bottle
Grade ACS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.