Learn More
Macherey-Nagel Resuspension Buffer RES‑EF, 1000 mL
Catalog No. NC1886448
Encompass_Preferred
Resuspension Buffer RES‑EF to be used with NucleoBond Xtra Midi / Maxi EF kits.
Catalog No. NC1886448 Supplier Macherey-Nagel Supplier No. 740386.1000
Resuspension Buffer RES‑EF to be used with NucleoBond Xtra Midi / Maxi EF kits.