Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Buffer Reagent for Aluminum in Brine, 100 mL

Supplier:  Hach Company 2310742

Encompass

Buffer Reagent Solution for aluminum in brine testing. 100mL.bottle

Catalog No. 50-199-5775


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.