Learn More
Rockland Immunochemicals 10X PBST (0.2 M Potassium Phosphate 1.5 M Sodium Chloride, 0.5% (v/v) Tween-20, pH 7.2) - MB-075-1000
SDP
Catalog No. NC0887817
Encompass_Preferred
Phosphate Buffer Saline-Tween is commonly use in washing procedure while performing western-blot, ELISA and, more generally immuno-assays.
Catalog No. NC0887817 Supplier Rockland Immunochemicals Supplier No. MB0751000
Phosphate Buffer Saline-Tween is commonly use in washing procedure while performing western-blot, ELISA and, more generally immuno-assays.