Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pH Buffer Salt, pH 10.4, in ready to mix packets, Certified, Fisher Chemical

Catalog No. B77 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
12 Packets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B77 12 Packets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B77

missing translation for 'validMainframeMsg'

Each packet contains sufficient material to prepare 1 liter of solution with a pH of 10.3 to 10.5 at 25°C

  • Each packet contains sufficient material to prepare 1 liter of solution with a pH of 10.4 ± 0.1 at 25°C
  • Convenient and ready-to-mix
  • Mix with deionized water to make 1L of pH buffer
  • Eliminates the need to store or carry bottles of buffer solution - simply transport the ready-to-mix packets
  • Intended for research use only; not for human or animal diagnostic purposes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Melting Point 168.5°C
Boiling Point 219°C
Color White
pH 10.4
Physical Form Powder or Solid
Quantity 12 Packets
Identification Pass Test
Packaging Envelopes
Grade Certified
Chemical Name or Material Buffer Salt, pH 10.5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.