Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer, Sodium Hypophosphite, pH 2.4, HPLC Mobile Phase for Analysis of Beverage Additives, Ricca Chemical
SDP

Catalog No. 14681 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1468-1 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1468-1 Supplier Ricca Chemical Company Supplier No. 14681
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HPLC mobile phase formulated for the analysis of beverage additives.

Specifications

Chemical Name or Material Buffer Sodium Hypophosphite
CAS 7732-18-5, 67-56-1, 67-63-0, 7664-38-2, 10039-56-2, 22767-49-3
CAS Min % 82.0, 11.57, 3.91, 0.12, 0.11, 0.09
CAS Max % 86.0, 12.05, 4.07, 0.12, 0.11, 0.1
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 2.4
Physical Form Liquid
Quantity 4 L
Solution Type pH Buffer
Packaging Amber Glass Bottle
Grade Not Available
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.