Learn More
Hach Company Buffer Solution, ULR Chlorine, pk/20
Supplier:  Hach Company 2493120
Encompass
For ultra-low Total Chlorine Determination by the DPD method. Hach Method 8370. Range: 2 - 500 μg/L Cl2 Ampuled for stability. Package of 20 1.5 mL ampules.*This product has not been evaluated to test for chlorine and chlormaines in medical applications in the United States.
Catalog No. 50-199-6072