Learn More
Hach Company Buffer Solution, pH 10.01, Color-coded Blue, 50 mL
Supplier: Hach Company 2283626
pH Buffer Standard Solution for pH electrode calibration. Color-coded pH buffer solution for easy identification. pH is 10.01 ±0.02 at 25°C. 50 mL pour-top bottle recommended for field use.