Learn More
Hach Company Buffer Solution, pH 10.01, Color-coded Blue, 4L
Supplier:  Hach Company 2283656
Encompass
pH Buffer Standard Solution for pH electrode calibration. Color-coded pH Buffer solution for easy identification. pH is 10.01 ±0.02 at 25°C. 4 L bottle
Catalog No. 50-199-5737