Learn More
Hach Company Hardness 1 Buffer Solution, 1 L
Supplier:  Hach Company 42453
Encompass
Buffer for hardness and chelant determination by titration, pH 10.1 ± 0.1 at 20C. Used with Hach methods 8213 and 8226. Also used with the Hach Phenols method 8047. 1 liter bottle.
Catalog No. 50-199-4283