Learn More
Hach Company Hardness 1 Buffer Solution, 100 mL MDB
Supplier:  Hach Company 42432
Encompass
Buffer for hardness and chelant determination by titration, pH 10.1 ± 0.1 at 20°C. Hach methods 8213 and 8226. Also used in Hach Phenols method 8047. 100 mL Marked Dropping Bottle
Catalog No. 50-199-4281