Learn More
Hach Company Hardness 1 Buffer Solution, 50mL SCDB
Supplier:  Hach Company 42426
Encompass
Buffer for hardness and chelant determination by titration, pH 10.1 ± 0.1 at 20C. For use with the Storm Water Test Kit (item #2481300,) and the Phenol Test Kit (item #2483600)
Catalog No. 50-199-4280