Learn More
Hach Company Buffer Solution, pH 4.01, 500 mL
Supplier:  Hach Company 1222349
Encompass
pH Buffer Standard Solution for pH electrode calibration. Colorless pH buffer solution. pH is 4.01 ±0.02 at 25°C. 500 mL bottle
Catalog No. 50-199-4863