Learn More
Hach Company Buffer Solution, pH 4.01, Color-coded Red, 20 L
Supplier:  Hach Company 2283461
Encompass
pH Buffer Standard Solution for pH electrode calibration. Color-coded pH buffer solution for easy identification. pH is 4.01 ±0.02 at 25°C. 20 L Bag-in-box package with a collapsible inner bladder in a white corrugated cardboard box. Ideal packaging size for laboratories doing high volume pH testing.
Catalog No. 50-199-5728