Learn More
Hach Company Buffer Solution, pH 4.01, Color-coded Red, 500 mL
Supplier:  Hach Company 2283449
Encompass
pH Buffer Standard Solution for pH electrode calibration. Color-coded pH Buffer solution for easy identification. pH is 4.01 ±0.02 at 25°C. 500 mL bottle
Catalog No. 50-199-5726