Learn More
Hach Company Buffer Solution, pH 7.00, Color-coded Yellow, 50 mL
Supplier:  Hach Company 2283526
Encompass
pH Buffer Standard Solution for pH electrode calibration. Color-coded pH buffer solution for easy identification. pH is 7.00 ±0.02 at 25° C. 50 mL pour-top bottle recommended for field use.
Catalog No. 50-199-5729