Learn More
Hach Company Buffer Solution, pH 7.00, Color-coded Yellow, 15 mL
Supplier:  Hach Company 2283536
Encompass
pH Buffer Standard Solution for pH electrode calibration. Color-coded pH buffer solution for easy identification. pH is 7.00 ±0.02 at 25°C. 15 mL bottle
Catalog No. 50-199-5730