Learn More
Hach Company Buffer Solution, pH 7.00, Color-coded Yellow, 500 mL
Supplier:  Hach Company 2283549
Encompass
pH Buffer Standard Solution for pH electrode calibration. Color-coded pH Buffer solution for easy identification. pH is 7.00 ±0.02 at 25°C. 500 mL bottle
Catalog No. 50-199-5731