Learn More
Hach Company Hach Buffer Solution for EZ4041 Total Hardness Analyzer, 2 L (APPC4041-02)
Catalog No. NC2227578 Shop All Hach Company Products
Encompass
Ready-to-use buffer solution formulated for routine calibration and maintenance of the EZ4041 Total Hardness Analyzer. Supplied in a 2 L bottle, this reagent eliminates the need for on-site preparation and helps maintain consistent total hardness measurements for online monitoring.
  • Ready-to-use liquid reagent for calibration and maintenance.
  • Supplied in a 2 L bottle for reduced replacement frequency.
  • Formulated to support accurate total hardness measurement.
  • Compatible with EZ4041 total hardness analyzers and EZ Series online analyzers.
  • Simplifies reagent handling and reduces preparation errors.
This item is not returnable. View return policy
Catalog No. NC2227578 Supplier Hach Company Supplier No. APPC404102
Ready-to-use buffer solution formulated for routine calibration and maintenance of the EZ4041 Total Hardness Analyzer. Supplied in a 2 L bottle, this reagent eliminates the need for on-site preparation and helps maintain consistent total hardness measurements for online monitoring.
  • Ready-to-use liquid reagent for calibration and maintenance.
  • Supplied in a 2 L bottle for reduced replacement frequency.
  • Formulated to support accurate total hardness measurement.
  • Compatible with EZ4041 total hardness analyzers and EZ Series online analyzers.
  • Simplifies reagent handling and reduces preparation errors.