Learn More
Buffer solution pH 4.00 (+/-0.022 @ 25oC) Colored Red Specpure NIST Traceable
Description
Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications. Buffer solutions are essential to maintain the suitable condition to keep the correct pH for enzymes in many organisms to work. It is also used in fermentation processes and in setting the correct conditions for dyes used in coloring fabrics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Buffer solution |
| CAS | 1310-73-2, 7778-77-0, 7732-18-5 |
| Color | Red |
| Physical Form | Liquid |
| Quantity | 4 L |
| MDL Number | MFCD00146206 |
| Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
| Solubility Information | Miscible with water. |
| Concentration or Composition (by Analyte or Components) | Sodium hydroxide: 0.7%; Potassium dihydrogen phosphate: 0.7%; water: 98.6% |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.