Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, Giordano, pH 6.4, for Wright's Stain, Certified, LabChem™
GSA_VA

Catalog No. p-5389144
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC125901 500 mL
LC125902 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC125901 Supplier LabChem, Inc. Supplier No. LC125901
Only null left
Add to Cart
Add to Cart

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Physical Form Liquid
Chemical Name or Material Buffer Solution, Giordano
Name Note pH 6.4, for Wright's Stain
Density 1g/mL
CAS Max % 98.975, 0.66, 0.32, 0.04, 0.005
CAS 7778-77-0, 7558-79-4, 50-00-0, 67-56-1
Packaging Poly Bottle
Solubility Soluble in water
Specific Gravity 1g/mL
Decomposition Information Carbon monoxide; Carbon dioxide
Color Colorless
pH 6.40 to 6.45
Quantity 500 mL
Linear Formula KH2PO4
Formula Weight 136.09
Grade Certified
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.