Learn More
Buffer solution pH 1.0 (20°C), Hydrochloric acid / potassium chloride, traceable to SRM from NIST and PTB, Solstice
Shop All Solstice Research Chemicals ProductsDescription
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Chemical Name or Material | Buffer Solution Ph 1.0 (20 °C) |
| CAS | 7732-18-5, 7647-01-0, 7447-40-7 |
| Quantity | 1 L |
| MDL Number | MFCD00146206 |
| UN Number | 1789 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.