Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer solution pH 1.0 (20°C), Hydrochloric acid / potassium chloride, traceable to SRM from NIST and PTB, Solstice

Catalog No. 60048355 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
6 x 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-355 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-355 Supplier Solstice Supplier No. 310446X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrochloric acid / potassium chloride, traceable to SRM from NIST and PTB

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution pH 1.0 (20°C)
Name Note Hydrochloric acid / potassium chloride, traceable to SRM from NIST and PTB
Boiling Point 100°C
CAS Min % ≥99.0000%
CAS 7647-01-0, 7447-40-7
CAS Max % ≤0.5000%
Density 1.000 g/cm3
Quantity 6 x 1 L
Solution Type Buffer
Linear Formula CH3CH2OH
MDL Number MFCD00003568
Packaging Poly Bottle
UN Number UN1789
Synonym Hydrochloric acid – potassium chloride buffer solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.