Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer solution pH 10.0 (20°C), Borax / sodium hydroxide, Solstice

Catalog No. 60046292 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
6 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-292 6 x 1 L
60-024-61 500 mL
60-024-60 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-046-292 Supplier Solstice Supplier No. 336496X1L
Add to Cart
Add to Cart

Borax / sodium hydroxide

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2 , 1330-43-4 , 7732-18-5

Specifications

Boiling Point 100°C
CAS Max % <0.1000%, ≤0.5000%
CAS Min % ≥0.2500%
Quantity 6 x 1 L
Solution Type Buffer
Packaging Poly Bottle
Chemical Name or Material Buffer solution pH 10.0 (20°C), Borax / sodium hydroxide
ShelfLife 1260 days from date of manufacture
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.