Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 10.00, MilliporeSigma™

Catalog No. MBX16361 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MBX16361 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MBX16361 Supplier MilliporeSigma Supplier No. BX16361

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Buffer Solution, pH 10.00
Physical Form Liquid
Quantity 500 mL
Linear Formula Mixture (Borate)
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.