Learn More
Specifications
Specifications
| Chemical Name or Material | Buffer Solution pH 11.0 at 25°C |
| Color | Colorless |
| CAS Max % | 99.64, 0.35, 0.01 |
| CAS | 7558-79-4, 1310-73-2 |
| Decomposition Information | Phosphine; Phosphorus oxides; Phosgene |
| Density | 1g/mL |
| pH | 11.00 ±0.05 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Identification | Passes Test |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.