Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution pH 12.0 at 25°C, LabChem™
GSA_VA

Catalog No. p-5379473
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC125601 500 mL
LC125602 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC125601 Supplier LabChem, Inc. Supplier No. LC125601
Only null left
Add to Cart
Add to Cart

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Chemical Name or Material Buffer Solution pH 12.0 at 25°C
Color Colorless
CAS Max % 99.58, 0.35, 0.07
CAS 7558-79-4, 1310-73-2
Decomposition Information Phosphine; Phosphorus oxides; Phosgene; Thermal decomposition generates: Corrosive vapors
Density 1g/mL
pH 12.00 ±0.05
Physical Form Liquid
Quantity 500 mL
Identification Passes Test
Linear Formula Na2HPO4
Packaging Poly Bottle
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 141.96
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.