Learn More
Buffer solution pH 13.0 (20°C), Glycine / sodium hydroxide / sodium chloride solution, Solstice
Shop All Solstice Research Chemicals ProductsDescription
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Chemical Name or Material | Buffer solution pH 13.0 (20°C) |
| Name Note | Glycine / sodium hydroxide / sodium chloride solution |
| CAS Min % | ≥90.0000% |
| CAS Max % | ≤100.0000%, <0.5000% |
| CAS | 1310-73-2, 6000-44-8, 7647-14-5 |
| Quantity | 6 x 1 L |
| Solution Type | Buffer |
| MDL Number | MFCD00146206 |
| Packaging | Poly Bottle |
| UN Number | UN1824 |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.