Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution pH 13.0 at 25°C, Certified, LabChem™

Catalog No. p-5379475
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer™
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC125651 500 mL Poly Bottle
LC125652 1 L Poly Bottle
LC125654 4 L Poly Bottle
LC125656 10 L Cubitainer™
LC125655 20 L Cubitainer™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC125651 Supplier LabChem, Inc. Supplier No. LC125651
Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Buffer Solution pH 13.0 at 25°C
Color Brown to Colorless
CAS Max % 99.24, 0.37, 0.34, 0.05
CAS 7447-40-7, 1310-73-2, 89-83-8
Decomposition Information Phosphine; Phosphorus oxides; Phosgene; Thermal decomposition generates: Corrosive vapors; Carbon dioxide; Carbon monoxide
Density 1g/mL
pH 13.00 ±0.05
Physical Form Liquid
Quantity 500 mL
Identification Passes Test
Linear Formula C10H14O
Packaging Poly Bottle
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 150.22
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.