Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 13.0, Reference Standard, Spectrum™ Chemical
SDP

Catalog No. 18601124 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
13.00 ±0.05
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-601-124 4 L
18-601-123 13.00 ±0.05
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-601-124 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B3784LTPL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

B-378

Spectrum™ Chemical Buffer Solution, pH 13.0, Reference Standard

Specifications

CAS 1310-73-2, 7732-18-5, 7747-40-7, 89-83-8
CAS Max % 100%
For Use With (Application) pH Measurement
For Use With (Equipment) pH Meter
pH 13.00 ±0.05
Physical Form Liquid
Quantity 4 L
Solution Type Buffer
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.