Learn More
Buffer solution, pH 2.00 (+/-0.026 @ 25oC), No Color, Specpure, NIST Traceable
Description
Buffer Solution pH 2 used as a means of keeping pH at a nearly constant value in a different chemical applications. Glycine Buffer Solution pH 2 used for eluting antibodies in various antibody purification procedures i.e. protein A-agarose, protein G-agarose, affinity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Buffer solution |
| CAS | 7447-40-7, 7647-01-0, 7732-18-5 |
| Color | Colorless |
| Density | 1.005g/cm3 at 20°C |
| Physical Form | Liquid |
| Quantity | 500 mL |
| MDL Number | MFCD00146252 |
| Traceability to NIST | Yes |
| Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
| Solubility Information | Soluble in water. |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.