Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer solution, pH 2.00 (+/-0.026 @ 25oC), No Color, Specpure, NIST Traceable

Catalog No. AA40447K2
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40447K2 1 L
AA40447AP 500 mL
AA40447K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA40447K2 Supplier Thermo Scientific Chemicals Supplier No. 040447K2
Only null left
Add to Cart

For Calibration of pH Meters | CAS: 7447-40-7

Buffer Solution pH 2 used as a means of keeping pH at a nearly constant value in a different chemical applications. Glycine Buffer Solution pH 2 used for eluting antibodies in various antibody purification procedures i.e. protein A-agarose, protein G-agarose, affinity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution
CAS 7447-40-7, 7647-01-0, 7732-18-5
Color Colorless
Density 1.005g/cm3 at 20°C
Physical Form Liquid
Quantity 1 L
MDL Number MFCD00146252
Traceability to NIST Yes
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Solubility Information Soluble in water.
Concentration or Composition (by Analyte or Components) Potassium chloride: 0.7%; Hydrochloric acid: 0.7%; water: 98.6%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.