Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 2.00 +/-0.02 at 25°C, Reference Standard, Spectrum™ Chemical
SDP

Catalog No. 18601127 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-127 20 L Poly Bottle
18-601-125 500 mL Poly Bottle
18-601-128 1 L Poly Bottle
18-601-126 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-127 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B22520LT
Only null left
Add to Cart
Add to Cart

B-225

Spectrum™ Chemical Buffer Solution, pH 2.00 ± 0.02 at 25°C, Reference Standard is a mixture of weak acid and its conjugate base in order to prevent changes in the pH of a solution. Buffer solutions let you calibrate any pH tester to ensure accurate readings when measuring pH in water samples and process liquids.

Specifications

CAS 7647-01-0, 7447-40-7, 7732-18-5
CAS Max % 1%, 98%
For Use With (Application) pH Measurement
For Use With (Equipment) pH Meter
pH 2.00 ±0.02
Physical Form Aqueous Solution
Quantity 20 L
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.