Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution pH 4.0 at 25°C, Certified, LabChem™
GSA_VA

Catalog No. LC122701 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer™
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC122701 500 mL Poly Bottle
LC122702 1 L Poly Bottle
LC122704 4 L Poly Bottle
LC122706 10 L Cubitainer™
LC122705 20 L Cubitainer™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC122701 Supplier LabChem, Inc. Supplier No. LC122701
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Weight: 204.23 g/mol

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Chemical Name or Material Buffer Solution pH 4.0 at 25°C
Color Colorless
CAS Max % 98.935, 1.02, 0.04, 0.005
CAS 877-24-7, 50-00-0, 67-56-1
Decomposition Information Formaldehyde; Carbon monoxide; Carbon dioxide
Density 1g/mL
For Use With (Application) pH Calibration
For Use With (Equipment) pH Meter
pH 4.00 to 4.01
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Identification Passes Test
Linear Formula KHC8H4O4
Packaging Poly Bottle
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 204.23
Grade Certified
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.