Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution pH 4.0 at 25°C, Certified, LabChem™

Catalog No. LC122706 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer™
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC122706 10 L Cubitainer™
LC122701 500 mL Poly Bottle
LC122702 1 L Poly Bottle
LC122704 4 L Poly Bottle
LC122705 20 L Cubitainer™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. LC122706 Supplier LabChem, Inc. Supplier No. LC122706
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemical Name or Material Buffer Solution pH 4.0 at 25°C
Color Colorless
CAS Max % 98.935, 1.02, 0.04, 0.005
CAS 877-24-7, 50-00-0, 67-56-1
Decomposition Information Formaldehyde; Carbon monoxide; Carbon dioxide
Density 1g/mL
For Use With (Application) pH Calibration
pH 4.00 to 4.01
Physical Form Liquid
Quantity 10 L
Solution Type pH Buffer
Identification Passes Test
Linear Formula KHC8H4O4
Packaging Cubitainer™
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 204.23
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.