Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 4.00, Color-Coded Red (Certified), Fisher Chemical™

Catalog No. p-213014 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Poly Bottle
PolyPac™
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SB101-500 500 mL Poly Bottle
SB101-4 4 L Poly Bottle
SB101-20 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. SB101-500
Only null left
Add to Cart
This item is not returnable. View return policy

pH/temperature conversions given on label

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer Solution, pH 4.00, Color-Coded Red
Absorbance Pass Test
Melting Point 0°C
CAS 50-00-0, 67-56-1, 877-24-7, 7732-18-5, 16423-68-0
Boiling Point 100°C
Color Red
Density 1.0g/cm³
Description pH/temperature conversions given on label.
pH 4.0
Physical Form Liquid
Quantity 500 mL
Identification Pass Test
MDL Number MFCD00146206
Packaging Poly Bottle
Specific Gravity 1
Traceability to NIST Traceable to NIST.
Solubility Information Soluble in water
Grade Certified
Concentration or Composition (by Analyte or Components) Potassium Hydrogen Phthalate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.