Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer solution pH 5, Acetate buffer

Catalog No. ac258620010
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC258620010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC258620010 Supplier Thermo Scientific Chemicals Supplier No. 258620010
Only null left
Add to Cart

CAS: 64-19-7

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

pH buffer
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution pH 5
CAS 64-19-7, 7732-18-5, 1310-58-3
Color Colorless
Density 1.0000g/mL
pH 4.98 to 5.02 (20°C)
Physical Form Liquid
Quantity 1 L
Formulation 0.7mL glacial acetic acid and 0.6g potassium hydroxide per liter
Packaging Plastic bottle
MDL Number MFCD03097943
Specific Gravity 1
Solubility Information Solubility in water: soluble.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.