Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer solution pH 6.865 (25°C), With fungicide, according to DIN 19266, Solstice

Catalog No. 60048368 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
6 x 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-368 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-368 Supplier Solstice Supplier No. 335946X1L
Add to Cart
Add to Cart

With fungicide, according to DIN 19266

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution pH 6.865 (25°C)
Name Note With fungicide, according to DIN 19266
Boiling Point 100°C
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%, ≤1.0000%
CAS 7558-79-4, 7778-77-0
Density 1.0 g/cm3
Quantity 6 x 1 L
Solution Type Buffer
Packaging Poly Bottle
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.