Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution pH 7.0 (20°C), Potassium Dihydrogen Phosphate / Disodium Hydrogen Phosphate, Traceable to SRM from NIST, with Fungicide, Honeywell Fluka™

Catalog No. 60046289 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
5 L
10 L
6 x 1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-289 6 x 1 L
60-024-52 500 mL
60-024-53 5 L
60-024-50 10 L
60-024-49 100 mL
60-024-51 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-046-289 Supplier Honeywell Chemicals Supplier No. 336466X1L
Only null left
Add to Cart
Add to Cart

Potassium dihydrogen phosphate / disodium hydrogen phosphate, traceable to SRM from NIST, with fungicide.

Leading the Industry For Over 65 Years. Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured.

Composition per liter:

  • 3.52 g Potassium dihydrogen phosphate (KH2PO4)
  • 7.26 g di-Sodium hydrogen phosphate-2-hydrate (Na2HPO4*2H20)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Synonym Potassium sodium phosphate buffer solution

Specifications

Boiling Point 100°C
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%, 0.7300%, 0.3500%
CAS 7558-79-4, 7778-77-0
Density 1 g/cm3
Quantity 6 x 1 L
Solution Type Buffer
MDL Number MFCD00146206
Packaging Poly Bottle
Chemical Name or Material Buffer solution pH 7.0 (20°C), Potassium dihydrogen phosphate / disodium hydrogen phosphate, traceable to SRM from NIST, with fungicide
ShelfLife 1260 days from date of manufacture
EINECSNumber 231-791-2
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.