Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 7.4, Reagents

Catalog No. 01337401
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-401 1 L
01-337-399 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-401 Supplier Reagents Holdings Llc Supplier No. C21560201A
Add to Cart
Add to Cart

Buffer Solution, pH 7.4, NIST Traceable, use for pH meter calibration. Manufactured in ISO 9001 facility. NIST Traceable.

Specifications

Chemical Name or Material Buffer Solution, pH 7.4
Melting Point 0°C
CAS 7732-18-5, 7778-77-0, 1310-73-2
Boiling Point 100°C
Color Colorless
For Use With (Application) Calibration
For Use With (Equipment) pH Meter
pH 7.4
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Poly Bottle
Specific Gravity 1.0048
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.