Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 7, Baker Analyzed, Yellow, J.T. Baker™

Catalog No. 14650318 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-650-318 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-650-318 Supplier Avantor Supplier No. 565601
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality buffer solution for accurate pH measurements.

Specifications

Chemical Name or Material Buffer, pH 7 Reference standard solution
Color Yellow
For Use With (Application) pH Calibration
pH 7
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Flash Point 0°C (32°F)
Packaging Poly
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.