Learn More
Aqua Solutions BUFFER SOLUTION PH 8.0 500ML
SDP
Catalog No. NC0951313
Encompass_Preferred
Buffer Reference Solution pH 8.00 0.02 @ 25C N.I.S.T. Traceable (500 ml)
Catalog No. NC0951313 Supplier Aqua Solutions Supplier No. 1510500ML
Buffer Reference Solution pH 8.00 0.02 @ 25C N.I.S.T. Traceable (500 ml)