Learn More
Hach Company Buffer Solution pH 9.23
Catalog No. 5000207004 Shop All Hach Company Products
Encompass
Buffer Solution-pH 9 23-500 mLpH Buffer Standard Solution for pH electrode calibration with Certificate of Analysis Colorless pH buffer pH is 9 23 0 02 at 25 C solution 500 mL bottle Available for download from uk hach com/CoA using product part number and lot number
Catalog No. 50-002-07004 Supplier Hach Company Supplier No. S11M011
Buffer Solution-pH 9 23-500 mLpH Buffer Standard Solution for pH electrode calibration with Certificate of Analysis Colorless pH buffer pH is 9 23 0 02 at 25 C solution 500 mL bottle Available for download from uk hach com/CoA using product part number and lot number