Learn More
Sigma Aldrich Fine Chemicals Biosciences pH 7.0 Buffer Solution | CAS N/A | MDL N/A | 500 mL
Catalog No. NC2580217
Encompass
"Buffer solution pH 7.0 | Purity: N/A | Mol Wt: N/A | CAS No: N/A | MDL No: N/A | 500 mL"This pH 7.0 buffer solution provides a neutral pH environment for various laboratory applications. It contains 500 mL of an aqueous buffer that resists changes in pH when acids or bases are added, allowing it to maintain a pH of 7.0. The solution is used to provide a stable, neutral pH for chemical reactions and processes requiring pH control.
Catalog No. NC2580217 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BX16351500ML
"Buffer solution pH 7.0 | Purity: N/A | Mol Wt: N/A | CAS No: N/A | MDL No: N/A | 500 mL"This pH 7.0 buffer solution provides a neutral pH environment for various laboratory applications. It contains 500 mL of an aqueous buffer that resists changes in pH when acids or bases are added, allowing it to maintain a pH of 7.0. The solution is used to provide a stable, neutral pH for chemical reactions and processes requiring pH control.